Learn More
Meso Scale Discovery U-PLEX Human MIF ASY384-well
SDP
Catalog No. 5000080718
Encompass
The Meso Scale Discovery U-PLEX Human MIF 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of macrophage migration inhibitory factor (MIF) in human serum plasma urine saliva CSF and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined workflow This 384-well assay allows high-throughput analysis of up to 80 samples per plate Each kit contains reagents for one 384-well plate including antibody-coated plates detection antibodies read buffer wash buffer diluent and calibrators
Catalog No. 50-000-80718 Supplier Meso Scale Discovery Supplier No. K251XJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIF 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of macrophage migration inhibitory factor (MIF) in human serum plasma urine saliva CSF and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined workflow This 384-well assay allows high-throughput analysis of up to 80 samples per plate Each kit contains reagents for one 384-well plate including antibody-coated plates detection antibodies read buffer wash buffer diluent and calibrators