Learn More
Meso Scale Discovery U-PLEX Human MIF ASYQP
SDP
Catalog No. 5000079789
Encompass
The U-PLEX Human MIF Assay (K151XJK-21) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of human Macrophage Migration Inhibitory Factor (MIF) in biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in research applications studying inflammation and immune responses
Catalog No. 50-000-79789 Supplier Meso Scale Discovery Supplier No. K151XJK21
The U-PLEX Human MIF Assay (K151XJK-21) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of human Macrophage Migration Inhibitory Factor (MIF) in biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in research applications studying inflammation and immune responses