Learn More
Meso Scale Discovery U-PLEX Human MIG ASY 25 PL
SDP
Catalog No. 5000080446
Encompass
The U-PLEX Human MIG Assay is a multiplex immunoassay kit that allows for the simultaneous measurement of human MIG in cell culture supernatants serum plasma (EDTA citrate heparin) and urine It uses electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR Imager 2400A from Meso Scale Discovery The assay can quantitatively measure human MIG concentrations with high sensitivity and broad dynamic range It includes a plate coated with capture antibodies detection antibodies labeled with ECL labels calibrators and controls The kit provides reagents for one 96-well plate and can test up to 40 samples in duplicate
Catalog No. 50-000-80446 Supplier Meso Scale Discovery Supplier No. K151AGYK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MIG Assay is a multiplex immunoassay kit that allows for the simultaneous measurement of human MIG in cell culture supernatants serum plasma (EDTA citrate heparin) and urine It uses electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR Imager 2400A from Meso Scale Discovery The assay can quantitatively measure human MIG concentrations with high sensitivity and broad dynamic range It includes a plate coated with capture antibodies detection antibodies labeled with ECL labels calibrators and controls The kit provides reagents for one 96-well plate and can test up to 40 samples in duplicate