Learn More
Meso Scale Discovery U-PLEX Human MIG ASY384-well
SDP
Catalog No. 5000081106
Encompass
The U-PLEX Human MIG Assay 384-well Kit from Meso Scale Discovery is a 384-well multiplex immunoassay kit for the quantitative measurement of human MIG in cell culture supernatants serum plasma and other biological fluids It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents It allows simultaneous measurement of human MIG concentrations in the range of 1 5-10 000 pg/mL
Catalog No. 50-000-81106 Supplier Meso Scale Discovery Supplier No. K251AGYK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MIG Assay 384-well Kit from Meso Scale Discovery is a 384-well multiplex immunoassay kit for the quantitative measurement of human MIG in cell culture supernatants serum plasma and other biological fluids It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents It allows simultaneous measurement of human MIG concentrations in the range of 1 5-10 000 pg/mL