Learn More
Meso Scale Discovery U-PLEX Human MIG ASYQP 25 PL
SDP
Catalog No. 5000079046
Encompass
The U-PLEX Human MIG Assay is a multiplex immunoassay kit for measuring human MIG in cell culture supernatants serum plasma (EDTA citrate heparin) and saliva It uses electrochemiluminescence detection and is compatible with the MESO QuickPlex SQ 120 The assay detects 0 03 - 1000 pg/mL and includes a plate with analyte-specific antibodies detection antibodies standards controls buffers substrate and consumables It requires low sample volume and offers high sensitivity reproducibility and recovery enabling efficient and accurate quantification of the chemokine MIG
Catalog No. 50-000-79046 Supplier Meso Scale Discovery Supplier No. K151AGYK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MIG Assay is a multiplex immunoassay kit for measuring human MIG in cell culture supernatants serum plasma (EDTA citrate heparin) and saliva It uses electrochemiluminescence detection and is compatible with the MESO QuickPlex SQ 120 The assay detects 0 03 - 1000 pg/mL and includes a plate with analyte-specific antibodies detection antibodies standards controls buffers substrate and consumables It requires low sample volume and offers high sensitivity reproducibility and recovery enabling efficient and accurate quantification of the chemokine MIG