Learn More
Meso Scale Discovery U-PLEX Human MIG ASYQP
SDP
Catalog No. 5000080468
Encompass
The U-PLEX Human MIG Assay is a multiplex immunoassay that measures the levels of human MIG in cell culture supernatants serum plasma and other biological fluids It uses electrochemiluminescence detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay simultaneously measures MIG in a single sample in under 3 hours It requires a small sample volume of 50 L and has a detection range of 45 - 100 000 pg/mL The assay provides reproducible and sensitive results comparable to ELISA It includes a plate coated with analyte-specific antibodies detection antibodies and calibrator and control solutions The assay is available in a kit format for 96 tests
Catalog No. 50-000-80468 Supplier Meso Scale Discovery Supplier No. K151AGYK22
The U-PLEX Human MIG Assay is a multiplex immunoassay that measures the levels of human MIG in cell culture supernatants serum plasma and other biological fluids It uses electrochemiluminescence detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay simultaneously measures MIG in a single sample in under 3 hours It requires a small sample volume of 50 L and has a detection range of 45 - 100 000 pg/mL The assay provides reproducible and sensitive results comparable to ELISA It includes a plate coated with analyte-specific antibodies detection antibodies and calibrator and control solutions The assay is available in a kit format for 96 tests