Learn More
Meso Scale Discovery U-PLEX Human MIG ASYQP
SDP
Catalog No. 5000080593
Encompass
The U-PLEX Human MIG Assay is a multiplex immunoassay kit that allows for the simultaneous measurement of human MIG in cell culture supernatants serum plasma (EDTA citrate heparin) and saliva It uses electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR Imager 2400A from Meso Scale Discovery The assay can quantify MIG levels from 15 6 to 1000 pg/mL with high sensitivity and broad dynamic range It includes a plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators and controls The kit provides reagents for one 96-well plate and can test 40 samples in duplicate It has high specificity and low cross-reactivity
Catalog No. 50-000-80593 Supplier Meso Scale Discovery Supplier No. K151AGYK21
The U-PLEX Human MIG Assay is a multiplex immunoassay kit that allows for the simultaneous measurement of human MIG in cell culture supernatants serum plasma (EDTA citrate heparin) and saliva It uses electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR Imager 2400A from Meso Scale Discovery The assay can quantify MIG levels from 15 6 to 1000 pg/mL with high sensitivity and broad dynamic range It includes a plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators and controls The kit provides reagents for one 96-well plate and can test 40 samples in duplicate It has high specificity and low cross-reactivity