Learn More
Meso Scale Discovery U-PLEX Human MIP-1 ASYQP
SDP
Catalog No. 5000079124
Encompass
The U-PLEX Human MIP-1 Assay (K151UJK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of MIP-1 (macrophage inflammatory protein-1 alpha) in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers Designed for research applications in inflammation immunology and drug development studies
Catalog No. 50-000-79124 Supplier Meso Scale Discovery Supplier No. K151UJK21
The U-PLEX Human MIP-1 Assay (K151UJK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of MIP-1 (macrophage inflammatory protein-1 alpha) in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers Designed for research applications in inflammation immunology and drug development studies