Learn More
Meso Scale Discovery U-PLEX Human MIP-1B ASY
SDP
Catalog No. 5000080533
Encompass
The Meso Scale Discovery U-PLEX Human MIP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human chemokine MIP-1 (CCL4) It uses electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be fully automated on the MSD instrument platform It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure MIP-1 in serum plasma cell culture supernatants and other biological sample types
Catalog No. 50-000-80533 Supplier Meso Scale Discovery Supplier No. K151UKK1
The Meso Scale Discovery U-PLEX Human MIP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of the human chemokine MIP-1 (CCL4) It uses electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be fully automated on the MSD instrument platform It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure MIP-1 in serum plasma cell culture supernatants and other biological sample types