Learn More
Meso Scale Discovery U-PLEX Human MIP-3 ASY
SDP
Catalog No. 5000079715
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3alpha Assay Kit quantifies human Macrophage Inflammatory Protein 3 alpha (MIP-3alpha) in various samples using electrochemiluminescent detection It features a pre-coated plate with capture antibodies followed by a detection antibody with an electrochemiluminescent label The kit includes reagents for 10 x 96-well plates measuring 80 samples in duplicate Sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL
Catalog No. 50-000-79715 Supplier Meso Scale Discovery Supplier No. K151UZK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human MIP-3alpha Assay Kit quantifies human Macrophage Inflammatory Protein 3 alpha (MIP-3alpha) in various samples using electrochemiluminescent detection It features a pre-coated plate with capture antibodies followed by a detection antibody with an electrochemiluminescent label The kit includes reagents for 10 x 96-well plates measuring 80 samples in duplicate Sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL