Learn More
Meso Scale Discovery U-PLEX Human MIP-3B ASY
SDP
Catalog No. 5000078767
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3beta Assay Kit is a multiplex immunoassay that measures human MIP-3beta in serum plasma cell culture supernatants and other biological samples It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify MIP-3beta concentrations as low as 45 pg/mL with as little as 25 L of sample It includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring samples The kit offers high performance immunoassays for biomarker research
Catalog No. 50-000-78767 Supplier Meso Scale Discovery Supplier No. K151VAK2
The Meso Scale Discovery U-PLEX Human MIP-3beta Assay Kit is a multiplex immunoassay that measures human MIP-3beta in serum plasma cell culture supernatants and other biological samples It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify MIP-3beta concentrations as low as 45 pg/mL with as little as 25 L of sample It includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring samples The kit offers high performance immunoassays for biomarker research