Learn More
Meso Scale Discovery U-PLEX Human MMP-1 ASY 25 PL
SDP
Catalog No. 5000079562
Encompass
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-1 protein in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The kit has a sensitivity of 0 03 ng/mL and an upper limit of quantification of 10 000 ng/mL for MMP-1
Catalog No. 50-000-79562 Supplier Meso Scale Discovery Supplier No. K151AHDK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-1 protein in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The kit has a sensitivity of 0 03 ng/mL and an upper limit of quantification of 10 000 ng/mL for MMP-1