Learn More
Meso Scale Discovery U-PLEX Human MMP-1 ASY
SDP
Catalog No. 5000080145
Encompass
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This product allows sensitive reproducible quantification of MMP-1 levels
Catalog No. 50-000-80145 Supplier Meso Scale Discovery Supplier No. K151AHDK1
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This product allows sensitive reproducible quantification of MMP-1 levels