Learn More
Meso Scale Discovery U-PLEX Human MMP-1 ASY384-well
SDP
Catalog No. 5000080998
Encompass
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a 384-well multiplex immunoassay kit that enables simultaneous detection and quantification of the human matrix metalloproteinase 1 (MMP-1) biomarker It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires a small sample volume and provides reproducible quantitative data from cell culture supernatants serum plasma and other biological matrices This product can aid research on MMP-1 involvement in various disease states
Catalog No. 50-000-80998 Supplier Meso Scale Discovery Supplier No. K251AHDK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a 384-well multiplex immunoassay kit that enables simultaneous detection and quantification of the human matrix metalloproteinase 1 (MMP-1) biomarker It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires a small sample volume and provides reproducible quantitative data from cell culture supernatants serum plasma and other biological matrices This product can aid research on MMP-1 involvement in various disease states