Learn More
Meso Scale Discovery U-PLEX Human MMP-1 ASYQP
SDP
Catalog No. 5000079561
Encompass
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantification of MMP-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The kit provides components for one 96-well plate and can test 40 samples in duplicate
Catalog No. 50-000-79561 Supplier Meso Scale Discovery Supplier No. K151AHDK22
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantification of MMP-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The kit provides components for one 96-well plate and can test 40 samples in duplicate