Learn More
Meso Scale Discovery U-PLEX Human MMP-1 ASYQP
SDP
Catalog No. 5000080492
Encompass
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is an electrochemiluminescent immunoassay for measuring MMP-1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The assay range is 0 03-400 ng/mL with no cross-reactivity with other MMPs It requires 50 L sample volume with results in under 3 hours The kit includes a 96-well plate capture and detection antibodies calibrator and diluent offering a simple reproducible method for quantifying MMP-1
Catalog No. 50-000-80492 Supplier Meso Scale Discovery Supplier No. K151AHDK21
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is an electrochemiluminescent immunoassay for measuring MMP-1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The assay range is 0 03-400 ng/mL with no cross-reactivity with other MMPs It requires 50 L sample volume with results in under 3 hours The kit includes a 96-well plate capture and detection antibodies calibrator and diluent offering a simple reproducible method for quantifying MMP-1