Learn More
Meso Scale Discovery U-PLEX Human MMP-2 ASY
SDP
Catalog No. 5000078402
Encompass
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-2 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-2 levels The assay has a detection range of 0 03-400 ng/mL and requires 25 L sample input It includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The total assay time is 3 5 hours
Catalog No. 50-000-78402 Supplier Meso Scale Discovery Supplier No. K151AHKK2
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-2 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-2 levels The assay has a detection range of 0 03-400 ng/mL and requires 25 L sample input It includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The total assay time is 3 5 hours