Learn More
Meso Scale Discovery U-PLEX Human MMP-2 ASY384-well
SDP
Catalog No. 5000080990
Encompass
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of MMP-2 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label calibrators and assay diluent The assay has a typical sensitivity of 0 04 ng/mL and an upper limit of quantification of 10 000 pg/mL It requires a low sample volume of 25 L and the total assay time is around 3 hours
Catalog No. 50-000-80990 Supplier Meso Scale Discovery Supplier No. K251AHKK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of MMP-2 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label calibrators and assay diluent The assay has a typical sensitivity of 0 04 ng/mL and an upper limit of quantification of 10 000 pg/mL It requires a low sample volume of 25 L and the total assay time is around 3 hours