Learn More
Meso Scale Discovery U-PLEX Human MMP-2 ASYQP
SDP
Catalog No. 5000081589
Encompass
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-2 in human serum plasma urine saliva CSF and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample consumption and rapid time to results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure MMP-2 in samples
Catalog No. 50-000-81589 Supplier Meso Scale Discovery Supplier No. K151AHKK22
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-2 in human serum plasma urine saliva CSF and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample consumption and rapid time to results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure MMP-2 in samples