Learn More
Meso Scale Discovery U-PLEX Human MMP-7 ASY 25 PL
SDP
Catalog No. 5000079049
Encompass
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-7 levels down to 0 04 ng/mL The assay requires a small sample volume of 50 L and has a 3 5 hour total assay time The U-PLEX format allows simultaneous measurement of MMP-7 with other biomarkers in the same sample This assay provides a flexible sensitive and efficient way to measure MMP-7 levels in biological samples
Catalog No. 50-000-79049 Supplier Meso Scale Discovery Supplier No. K151AHEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-7 levels down to 0 04 ng/mL The assay requires a small sample volume of 50 L and has a 3 5 hour total assay time The U-PLEX format allows simultaneous measurement of MMP-7 with other biomarkers in the same sample This assay provides a flexible sensitive and efficient way to measure MMP-7 levels in biological samples