Learn More
Meso Scale Discovery U-PLEX Human MMP-7 ASY384-well
SDP
Catalog No. 5000078379
Encompass
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is a 384-well electrochemiluminescent immunoassay designed to measure levels of active and pro-MMP-7 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 384-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay has a typical sensitivity of 0 03 ng/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-78379 Supplier Meso Scale Discovery Supplier No. K251AHEK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is a 384-well electrochemiluminescent immunoassay designed to measure levels of active and pro-MMP-7 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 384-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay has a typical sensitivity of 0 03 ng/mL and an upper limit of quantification of 2500 pg/mL