Learn More
Meso Scale Discovery U-PLEX HUMAN MMP-9 ASSAY, Assay Format: 96-well plate, Dynamic Range: 1.6 - 10,000 pg/mL, Sensitivity: 1.6 pg/mL, Sample Types: EDTA Plasma, Serum, Tumor Lysate, Not for Diagnostic Use
SDP
Catalog No. NC3036724
Encompass
U-PLEX HUMAN MMP-9 ASSAY, Assay Format: 96-well plate, Dynamic Range: 1.6 - 10,000 pg/mL, Sensitivity: 1.6 pg/mL, Sample Types: EDTA Plasma, Serum, Tumor Lysate, Not for Diagnostic Use
Catalog No. NC3036724 Supplier Meso Scale Discovery Supplier No. K151AHMK2
U-PLEX HUMAN MMP-9 ASSAY, Assay Format: 96-well plate, Dynamic Range: 1.6 - 10,000 pg/mL, Sensitivity: 1.6 pg/mL, Sample Types: EDTA Plasma, Serum, Tumor Lysate, Not for Diagnostic Use