Learn More
Meso Scale Discovery U-PLEX Human MMP-9 total ASY
SDP
Catalog No. 5000078805
Encompass
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-9 levels down to 0 04 ng/mL The assay requires a small sample volume of 50 L and has a 3 5 hour total assay time The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrator and sample diluent It provides a simple reproducible method to quantify MMP-9 levels for research use
Catalog No. 50-000-78805 Supplier Meso Scale Discovery Supplier No. K151AHMK1
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-9 levels down to 0 04 ng/mL The assay requires a small sample volume of 50 L and has a 3 5 hour total assay time The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrator and sample diluent It provides a simple reproducible method to quantify MMP-9 levels for research use