Learn More
Meso Scale Discovery U-PLEX Human Nectin-4 ASY
SDP
Catalog No. 5000081587
Encompass
The Meso Scale Discovery Human Nectin-4 Singleplex Assay Kit is a singleplex immunoassay that measures Nectin-4 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity wide dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The assay has a typical lower limit of detection of 0 03 ng/mL and an upper limit of quantification of 2500 ng/mL This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control
Catalog No. 50-000-81587 Supplier Meso Scale Discovery Supplier No. K151AHFK1
The Meso Scale Discovery Human Nectin-4 Singleplex Assay Kit is a singleplex immunoassay that measures Nectin-4 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity wide dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The assay has a typical lower limit of detection of 0 03 ng/mL and an upper limit of quantification of 2500 ng/mL This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control