Learn More
Meso Scale Discovery U-PLEX Human Nectin-4 ASYQP
SDP
Catalog No. 5000080596
Encompass
The Meso Scale Discovery U-PLEX Human Nectin-4 Assay is a multiplex immunoassay kit that enables simultaneous quantification of Nectin-4 in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve Results are obtained in as little as 3 5 hours using an MSD instrument This assay is for research use only
Catalog No. 50-000-80596 Supplier Meso Scale Discovery Supplier No. K151AHFK21
The Meso Scale Discovery U-PLEX Human Nectin-4 Assay is a multiplex immunoassay kit that enables simultaneous quantification of Nectin-4 in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve Results are obtained in as little as 3 5 hours using an MSD instrument This assay is for research use only