Learn More
Meso Scale Discovery U-PLEX Human NGAL/LCN2 ASY
SDP
Catalog No. 5000078833
Encompass
The Meso Scale Discovery U-PLEX Human NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in human serum plasma and urine This assay uses MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 78 - 50 000 pg/mL and shows no cross-reactivity with other human proteins It requires a low sample volume of 50 L and provides results in under 3 hours This assay can aid in the early detection of acute kidney injury
Catalog No. 50-000-78833 Supplier Meso Scale Discovery Supplier No. K151Z1K1
The Meso Scale Discovery U-PLEX Human NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in human serum plasma and urine This assay uses MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 78 - 50 000 pg/mL and shows no cross-reactivity with other human proteins It requires a low sample volume of 50 L and provides results in under 3 hours This assay can aid in the early detection of acute kidney injury