Learn More
Meso Scale Discovery U-PLEX Human NGAL/LCN2 ASYQP
SDP
Catalog No. 5000078834
Encompass
The Meso Scale Discovery U-PLEX Human NGAL (Lipocalin-2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human neutrophil gelatinase-associated lipocalin (NGAL) in serum plasma urine cell culture supernatants and other biological samples This assay utilizes MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 15 6-2000 pg/mL and shows no cross-reactivity with other human lipocalins The assay requires a small sample volume of 50 L and provides results in under 3 hours This product can aid in the study of acute kidney injury and other inflammatory conditions where NGAL is upregulated
Catalog No. 50-000-78834 Supplier Meso Scale Discovery Supplier No. K151Z1K22
The Meso Scale Discovery U-PLEX Human NGAL (Lipocalin-2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human neutrophil gelatinase-associated lipocalin (NGAL) in serum plasma urine cell culture supernatants and other biological samples This assay utilizes MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 15 6-2000 pg/mL and shows no cross-reactivity with other human lipocalins The assay requires a small sample volume of 50 L and provides results in under 3 hours This product can aid in the study of acute kidney injury and other inflammatory conditions where NGAL is upregulated