Learn More
Meso Scale Discovery U-PLEX Human OX40/TNFRSF4 ASY
SDP
Catalog No. 5000081302
Encompass
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay for measuring OX40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and diluents It detects OX40 from 17 pg/mL to 100 000 pg/mL in 3 5 hours enabling quantification of the OX40 biomarker related to T cell activation
Catalog No. 50-000-81302 Supplier Meso Scale Discovery Supplier No. K151T7K1
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay for measuring OX40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and diluents It detects OX40 from 17 pg/mL to 100 000 pg/mL in 3 5 hours enabling quantification of the OX40 biomarker related to T cell activation