Learn More
Meso Scale Discovery U-PLEX Human P-Selectin ASYQP
SDP
Catalog No. 5000078807
Encompass
The Meso Scale Discovery U-PLEX Human P-Selectin Assay is a multiplex immunoassay kit for measuring P-selectin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L or less sample volume it is compatible with MSD instruments The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables sensitive reproducible quantification of P-selectin a key biomarker of platelet activation
Catalog No. 50-000-78807 Supplier Meso Scale Discovery Supplier No. K151AHNK21
The Meso Scale Discovery U-PLEX Human P-Selectin Assay is a multiplex immunoassay kit for measuring P-selectin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L or less sample volume it is compatible with MSD instruments The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables sensitive reproducible quantification of P-selectin a key biomarker of platelet activation