Learn More
Meso Scale Discovery U-PLEX Human PD-L2 ASY 25 PL
SDP
Catalog No. 5000079581
Encompass
The Meso Scale Discovery U-PLEX Human PD-L2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Programmed Death Ligand 2 (PD-L2) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor endogenous levels of human PD-L2 or changes after stimulation and provides a tool for understanding PD-L2 biology and evaluating checkpoint inhibitor therapies targeting the PD-1/PD-L2 pathway
Catalog No. 50-000-79581 Supplier Meso Scale Discovery Supplier No. K151A8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Programmed Death Ligand 2 (PD-L2) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor endogenous levels of human PD-L2 or changes after stimulation and provides a tool for understanding PD-L2 biology and evaluating checkpoint inhibitor therapies targeting the PD-1/PD-L2 pathway