Learn More
Meso Scale Discovery U-PLEX Human PD-L2 ASY384-well
SDP
Catalog No. 5000081000
Encompass
The Meso Scale Discovery U-PLEX Human PD-L2 384-Well Assay is a multiplex immunoassay panel that measures the PD-L2 protein in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of PD-L2 levels with as little as 25 L of sample It can be used to monitor immune response and has applications in immunotherapy and vaccine development research With a short 3 5 hour protocol this assay enables high-throughput analysis of PD-L2
Catalog No. 50-000-81000 Supplier Meso Scale Discovery Supplier No. K251A8K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD-L2 384-Well Assay is a multiplex immunoassay panel that measures the PD-L2 protein in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of PD-L2 levels with as little as 25 L of sample It can be used to monitor immune response and has applications in immunotherapy and vaccine development research With a short 3 5 hour protocol this assay enables high-throughput analysis of PD-L2