Learn More
Meso Scale Discovery U-PLEX Human PD-L2 ASYQP
SDP
Catalog No. 5000080385
Encompass
The Meso Scale Discovery U-PLEX Human PD-L2 Assay is a multiplex immunoassay kit for measuring human PD-L2 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables rapid and reliable quantification of PD-L2 crucial for regulating T cell activation and immune responses
Catalog No. 50-000-80385 Supplier Meso Scale Discovery Supplier No. K151A8K21
The Meso Scale Discovery U-PLEX Human PD-L2 Assay is a multiplex immunoassay kit for measuring human PD-L2 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables rapid and reliable quantification of PD-L2 crucial for regulating T cell activation and immune responses