Learn More
Meso Scale Discovery U-PLEX Human Pentraxin 3 ASY
SDP
Catalog No. 5000078398
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay kit for measuring Pentraxin 3 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 7-log dynamic range and low sample volume needs This assay quantifies Pentraxin 3 an inflammatory biomarker aiding in understanding immune function and inflammation The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X diluent and readout buffer for analyzing 48-96 samples
Catalog No. 50-000-78398 Supplier Meso Scale Discovery Supplier No. K151AGZK2
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay kit for measuring Pentraxin 3 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 7-log dynamic range and low sample volume needs This assay quantifies Pentraxin 3 an inflammatory biomarker aiding in understanding immune function and inflammation The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X diluent and readout buffer for analyzing 48-96 samples