Learn More
Meso Scale Discovery U-PLEX Human Pentraxin 3 ASY
SDP
Catalog No. 5000079519
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Pentraxin 3 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 2400 or SECTOR S 600 multi-array plate readers with integrated fluidics The assay has a detection range of 0 01 - 1000 ng/mL and includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators The total assay time is 3 5 hours This quantitative immunoassay provides sensitive and accurate Pentraxin 3 quantification for life science research
Catalog No. 50-000-79519 Supplier Meso Scale Discovery Supplier No. K151AGZK1
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Pentraxin 3 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 2400 or SECTOR S 600 multi-array plate readers with integrated fluidics The assay has a detection range of 0 01 - 1000 ng/mL and includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators The total assay time is 3 5 hours This quantitative immunoassay provides sensitive and accurate Pentraxin 3 quantification for life science research