Learn More
Meso Scale Discovery U-PLEX Human Pentraxin 3 ASYQP
SDP
Catalog No. 5000079044
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Pentraxin 3 and other biomarkers in human serum plasma and cell culture supernatants The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay requires a Meso Scale Discovery instrument and plates to run It includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls The kit provides reagents for one 96-well plate and can test up to 40 samples in duplicate
Catalog No. 50-000-79044 Supplier Meso Scale Discovery Supplier No. K151AGZK21
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Pentraxin 3 and other biomarkers in human serum plasma and cell culture supernatants The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay requires a Meso Scale Discovery instrument and plates to run It includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls The kit provides reagents for one 96-well plate and can test up to 40 samples in duplicate