Learn More
Meso Scale Discovery U-PLEX Human Pentraxin 3 ASYQP
SDP
Catalog No. 5000081648
Encompass
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is an electrochemiluminescent immunoassay for measuring Pentraxin 3 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the MESO QuickPlex SQ 120 The detection range is 0 04 to 10 000 ng/mL The assay aids in diagnosing inflammatory conditions and cardiovascular disease The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and sample diluent
Catalog No. 50-000-81648 Supplier Meso Scale Discovery Supplier No. K151AGZK22
The Meso Scale Discovery U-PLEX Human Pentraxin 3 Assay is an electrochemiluminescent immunoassay for measuring Pentraxin 3 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the MESO QuickPlex SQ 120 The detection range is 0 04 to 10 000 ng/mL The assay aids in diagnosing inflammatory conditions and cardiovascular disease The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and sample diluent