Learn More
Meso Scale Discovery U-PLEX Human Perforin ASY
SDP
Catalog No. 5000081908
Encompass
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures the levels of perforin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the simultaneous measurement of perforin along with other biomarkers in the same sample to understand immune response It requires 50 L of sample and can perform up to 80 tests per kit The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls
Catalog No. 50-000-81908 Supplier Meso Scale Discovery Supplier No. K151AHGK1
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures the levels of perforin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the simultaneous measurement of perforin along with other biomarkers in the same sample to understand immune response It requires 50 L of sample and can perform up to 80 tests per kit The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls