Learn More
Meso Scale Discovery U-PLEX Human Perforin ASYQP
SDP
Catalog No. 5000079563
Encompass
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures human Perforin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows simultaneous measurement of Perforin along with other biomarkers It requires 50 L or less of sample and can be read on MSD instrument platforms The U-PLEX format provides rapid results in just 3 5 hours This assay is for research use only not for use in diagnostic procedures
Catalog No. 50-000-79563 Supplier Meso Scale Discovery Supplier No. K151AHGK21
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures human Perforin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows simultaneous measurement of Perforin along with other biomarkers It requires 50 L or less of sample and can be read on MSD instrument platforms The U-PLEX format provides rapid results in just 3 5 hours This assay is for research use only not for use in diagnostic procedures