Learn More
Meso Scale Discovery U-PLEX Human Perforin ASYQP
SDP
Catalog No. 5000079564
Encompass
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures human Perforin in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies calibrators and buffers The assay has high sensitivity and wide dynamic range requires low sample volume and provides results in as little as 3 5 hours This product allows for the accurate quantification of Perforin which plays a key role in immune cell-mediated cytotoxicity
Catalog No. 50-000-79564 Supplier Meso Scale Discovery Supplier No. K151AHGK22
The Meso Scale Discovery U-PLEX Human Perforin Assay is a multiplex immunoassay that measures human Perforin in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies calibrators and buffers The assay has high sensitivity and wide dynamic range requires low sample volume and provides results in as little as 3 5 hours This product allows for the accurate quantification of Perforin which plays a key role in immune cell-mediated cytotoxicity