Learn More
Meso Scale Discovery U-PLEX Human PlGF ASYQP
SDP
Catalog No. 5000078328
Encompass
The Meso Scale Discovery U-PLEX Human PIGF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Placental Growth Factor (PIGF) in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid time-to-result This assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ120 or QuickPlex SQ120 Each kit provides reagents for 96 tests in a microplate format
Catalog No. 50-000-78328 Supplier Meso Scale Discovery Supplier No. K151B8K22
The Meso Scale Discovery U-PLEX Human PIGF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Placental Growth Factor (PIGF) in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid time-to-result This assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ120 or QuickPlex SQ120 Each kit provides reagents for 96 tests in a microplate format