Learn More
Meso Scale Discovery U-PLEX Human PP ASY384-well
SDP
Catalog No. 5000080229
Encompass
The Meso Scale Discovery U-PLEX Human PP 384-Well Assay is a multiplex immunoassay for quantifying 384 protein biomarkers in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection and supports automated liquid handling The panel includes cytokines chemokines growth factors and more Requiring only 25 L the 384-well format allows high-throughput analysis The assay completes in about 3 hours with sensitivity in the pg/mL range streamlining large-scale protein biomarker profiling
Catalog No. 50-000-80229 Supplier Meso Scale Discovery Supplier No. K2516AK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PP 384-Well Assay is a multiplex immunoassay for quantifying 384 protein biomarkers in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection and supports automated liquid handling The panel includes cytokines chemokines growth factors and more Requiring only 25 L the 384-well format allows high-throughput analysis The assay completes in about 3 hours with sensitivity in the pg/mL range streamlining large-scale protein biomarker profiling