Learn More
Meso Scale Discovery U-PLEX Human PP ASYQP 25 PL
SDP
Catalog No. 5000081690
Encompass
The U-PLEX Human PP Assay (K1516AK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of pancreatic polypeptide (PP) in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT technology for sensitive and specific detection The kit includes pre-coated plates detection antibodies calibrators and controls for reliable results Suitable for research applications in metabolic studies and diabetes research
Catalog No. 50-000-81690 Supplier Meso Scale Discovery Supplier No. K1516AK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human PP Assay (K1516AK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of pancreatic polypeptide (PP) in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT technology for sensitive and specific detection The kit includes pre-coated plates detection antibodies calibrators and controls for reliable results Suitable for research applications in metabolic studies and diabetes research