Learn More
Meso Scale Discovery U-PLEX Human Proinsulin ASYQP
SDP
Catalog No. 5000078905
Encompass
The MSD U-PLEX Human Proinsulin Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of proinsulin levels in human biological samples This singleplex format allows focused analysis of proinsulin a key biomarker for pancreatic beta-cell function and diabetes research The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods
Catalog No. 50-000-78905 Supplier Meso Scale Discovery Supplier No. K1516MK21
The MSD U-PLEX Human Proinsulin Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of proinsulin levels in human biological samples This singleplex format allows focused analysis of proinsulin a key biomarker for pancreatic beta-cell function and diabetes research The U-PLEX platform offers superior sensitivity wide dynamic range and minimal sample volume requirements compared to traditional ELISA methods