Learn More
Meso Scale Discovery U-PLEX Human proMMP-9 ASY
SDP
Catalog No. 5000080473
Encompass
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a multiplex immunoassay that measures ProMMP-9 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours It includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound an MSD read buffer a diluent and calibrators and controls This allows for the accurate and reliable quantification of ProMMP-9 levels in biological samples
Catalog No. 50-000-80473 Supplier Meso Scale Discovery Supplier No. K151AHLK1
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a multiplex immunoassay that measures ProMMP-9 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours It includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound an MSD read buffer a diluent and calibrators and controls This allows for the accurate and reliable quantification of ProMMP-9 levels in biological samples