Learn More
Meso Scale Discovery U-PLEX Human RANTES ASY
SDP
Catalog No. 5000079513
Encompass
The Meso Scale Discovery U-PLEX Human RANTES Assay is a multiplex immunoassay that measures RANTES levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Instruments The lower limit of detection is 0 04 pg/mL This assay allows for the quantitative measurement of RANTES which plays a role in immune system regulation
Catalog No. 50-000-79513 Supplier Meso Scale Discovery Supplier No. K151A2K1
The Meso Scale Discovery U-PLEX Human RANTES Assay is a multiplex immunoassay that measures RANTES levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Instruments The lower limit of detection is 0 04 pg/mL This assay allows for the quantitative measurement of RANTES which plays a role in immune system regulation