Learn More
Meso Scale Discovery U-PLEX Human RBP4 ASY 25 PL
SDP
Catalog No. 5000079079
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for measuring retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible detection with a dynamic range and low sample volume The detection limit is 17 pg/mL and the quantification limit is 10 000 pg/mL The kit includes plates with capture and detection antibodies calibrator and diluent for flexible rapid testing ensuring accurate RBP4 measurement for research
Catalog No. 50-000-79079 Supplier Meso Scale Discovery Supplier No. K151V9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for measuring retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible detection with a dynamic range and low sample volume The detection limit is 17 pg/mL and the quantification limit is 10 000 pg/mL The kit includes plates with capture and detection antibodies calibrator and diluent for flexible rapid testing ensuring accurate RBP4 measurement for research