Learn More
Meso Scale Discovery U-PLEX Human RBP4 ASY
SDP
Catalog No. 5000078303
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible detection of RBP4 levels with high specificity and low background The assay requires a small sample volume and provides a rapid 3 5 hour protocol The U-PLEX RBP4 Assay provides accurate quantitation of RBP4 levels which can aid in the study of insulin resistance diabetes and metabolic syndrome
Catalog No. 50-000-78303 Supplier Meso Scale Discovery Supplier No. K151V9K1
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible detection of RBP4 levels with high specificity and low background The assay requires a small sample volume and provides a rapid 3 5 hour protocol The U-PLEX RBP4 Assay provides accurate quantitation of RBP4 levels which can aid in the study of insulin resistance diabetes and metabolic syndrome