Learn More
Meso Scale Discovery U-PLEX Human RBP4 ASY
SDP
Catalog No. 5000080499
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of RBP4 levels with high specificity and low background The assay requires a small sample volume and provides a rapid 3 5 hour protocol The U-PLEX platform provides researchers with an efficient and robust tool for investigating RBP4 biology and biomarker applications
Catalog No. 50-000-80499 Supplier Meso Scale Discovery Supplier No. K151V9K2
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of RBP4 levels with high specificity and low background The assay requires a small sample volume and provides a rapid 3 5 hour protocol The U-PLEX platform provides researchers with an efficient and robust tool for investigating RBP4 biology and biomarker applications