Learn More
Meso Scale Discovery U-PLEX Human RBP4 ASY384-well
SDP
Catalog No. 5000080651
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 384-Well Assay Kit is a multiplex immunoassay that measures RBP4 levels in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of RBP4 The assay has high sensitivity wide dynamic range and requires low sample volume This product can help researchers studying RBP4 biology and its role in diseases like diabetes obesity and metabolic syndrome
Catalog No. 50-000-80651 Supplier Meso Scale Discovery Supplier No. K251V9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RBP4 384-Well Assay Kit is a multiplex immunoassay that measures RBP4 levels in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of RBP4 The assay has high sensitivity wide dynamic range and requires low sample volume This product can help researchers studying RBP4 biology and its role in diseases like diabetes obesity and metabolic syndrome