Learn More
Meso Scale Discovery U-PLEX Human RBP4 ASYQP
SDP
Catalog No. 5000078978
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for measuring retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible detection with a dynamic range and low sample volume Detection limits are 17 pg/mL to 50 000 pg/mL The assay takes about 3 5 hours for 96 tests using an MSD instrument offering a rapid accurate method for quantifying RBP4 in research and drug development
Catalog No. 50-000-78978 Supplier Meso Scale Discovery Supplier No. K151V9K21
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for measuring retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible detection with a dynamic range and low sample volume Detection limits are 17 pg/mL to 50 000 pg/mL The assay takes about 3 5 hours for 96 tests using an MSD instrument offering a rapid accurate method for quantifying RBP4 in research and drug development